Skip to Content
MilliporeSigma
2-(2H-Benzotriazol-2-yl)-4,6-bis(1-methyl-1-phenylethyl)phenol analytical standard

2-(2H-Benzotriazol-2-yl)-4,6-bis(1-methyl-1-phenylethyl)phenol

2-(2H-Benzotriazol-2-yl)-4,6-bis(1-methyl-1-phenylethyl)phenol analytical standard
Empirical Formula (Hill Notation):
C30H29N3O
CAS Number:
Molecular Weight:
447.57

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Format
Quality Segment
Markush Class
Markush Group
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing