Skip to Content
MilliporeSigma
(2,2-Dimethyl-3-methylsulfonyloxypropyl) methanesulfonate analytical standard

(2,2-Dimethyl-3-methylsulfonyloxypropyl) methanesulfonate

(2,2-Dimethyl-3-methylsulfonyloxypropyl) methanesulfonate analytical standard
Synonym(s):
Neopentyl glycol dimethylsulfate, 2,2-Dimethyl-1,3-propanediol dimethylsulfate
Empirical Formula (Hill Notation):
C7H16O6S2
CAS Number:
Molecular Weight:
260.33

Products

Brand
Formula Weight
Melting Point (°C)
Format
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing