Skip to Content
MilliporeSigma
2,2-Bis(hydroxymethyl)propionic acid 98%

2,2-Bis(hydroxymethyl)propionic acid

2,2-Bis(hydroxymethyl)propionic acid 98%
Synonym(s):
Bis-MPA, DMPA®, Dimethylolpropionic acid, 2,2-Bis(hydroxymethyl)propionic acid
Linear Formula:
(HOCH2)2C(CH3)COOH
CAS Number:
Molecular Weight:
134.13

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing