Skip to Content
MilliporeSigma
2,2′,3,4,4′,5′-Hexachlorobiphenyl (IUPAC No. 138) BCR®, certified reference material

2,2′,3,4,4′,5′-Hexachlorobiphenyl (IUPAC No. 138)

2,2′,3,4,4′,5′-Hexachlorobiphenyl (IUPAC No. 138) BCR®, certified reference material
Synonym(s):
2,2′,3,4,4′,5′-Hexachlorobiphenyl
Empirical Formula (Hill Notation):
C12H4Cl6
CAS Number:
Molecular Weight:
360.88

Products

Formula Weight
Manufacturer
Agency
Format
Reference Material Type
Special Grade
Showing 1-1 of 1