Skip to Content
MilliporeSigma
2,2,3,3-Tetrafluoropropyl methacrylate 99%

2,2,3,3-Tetrafluoropropyl methacrylate

2,2,3,3-Tetrafluoropropyl methacrylate 99%
Linear Formula:
CH2=C(CH3)COOCH2CF2CF2H
CAS Number:
Molecular Weight:
200.13

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing