Skip to Content
MilliporeSigma
2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol 98%

2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol

2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol 98%
Linear Formula:
HOCH2(CF2)4CH2OH
CAS Number:
Molecular Weight:
262.10

Products

Product Category
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing