Skip to Content
MilliporeSigma
vibrant-m

21-Acetoxy-17-α-hydroxypregna-1,4-diene-3,11,20-trione

vibrant-m
Synonym(s):
17-Hydroxy-3,11,20-trioxopregna-1,4-dien-21-yl acetate, Prednisone acetate
Empirical Formula (Hill Notation):
C23H28O6
CAS Number:
Molecular Weight:
400.46

Products

Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Special Grade