Skip to Content
MilliporeSigma
(20S)-20-Hydroxypregn-4-en-3-one Pharmaceutical Secondary Standard; Certified Reference Material

(20S)-20-Hydroxypregn-4-en-3-one

(20S)-20-Hydroxypregn-4-en-3-one Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
4-Pregnen-20α-ol-3-one, 20α-Hydroxy-4-pregnen-3-one, 20α-Hydroxyprogesterone
Empirical Formula (Hill Notation):
C21H32O2
CAS Number:
Molecular Weight:
316.48

Products

Formula Weight
Physical Form
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1