Skip to Content
MilliporeSigma
1-(Trimethylsiloxy)cyclopentene 97%

1-(Trimethylsiloxy)cyclopentene

1-(Trimethylsiloxy)cyclopentene 97%
Synonym(s):
1-Cyclopentenyloxytrimethylsilane, Cyclopentanone enol trimethyl silyl ether
Linear Formula:
C5H7OSi(CH3)3
CAS Number:
Molecular Weight:
156.30
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing