Skip to Content
MilliporeSigma
(1S,4S,8S)-5-Benzyl-2-isobutyl-8-methoxy-1,8-dimethylbicyclo[2.2.2]octa-2,5-diene 97%

(1S,4S,8S)-5-Benzyl-2-isobutyl-8-methoxy-1,8-dimethylbicyclo[2.2.2]octa-2,5-diene

(1S,4S,8S)-5-Benzyl-2-isobutyl-8-methoxy-1,8-dimethylbicyclo[2.2.2]octa-2,5-diene 97%
Synonym(s):
(1S,4S,8S) Carreira DOLEFIN Ligand, (1S,4S,8S)--5-Benzyl-8-methoxy-1,8-dimethyl-2-(2′-methylpropyl)-bicyclo[2.2.2]octa-2,5-diene, (1S,4S,8S)-5-Benzyl-2-isobutyl-8-methoxy-1,8-dimethyl-2-bicyclo[2.2.2]octa-2,5-diene
Empirical Formula (Hill Notation):
C22H30O
CAS Number:
Molecular Weight:
310.47

Products

Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1