Skip to Content
MilliporeSigma
(1S,2S)-(+)-1,2-Diaminocyclohexane 98%

(1S,2S)-(+)-1,2-Diaminocyclohexane

(1S,2S)-(+)-1,2-Diaminocyclohexane 98%
Synonym(s):
(1S)-trans-1,2-Cyclohexanediamine, (1S)-trans-1,2-Diaminocyclohexane
Linear Formula:
C6H10(NH2)2
CAS Number:
Molecular Weight:
114.19

Products

Product Category
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing