Skip to Content
MilliporeSigma
(1S,2S,3R,5S)-(+)-Pinanediol 99%

(1S,2S,3R,5S)-(+)-Pinanediol

(1S,2S,3R,5S)-(+)-Pinanediol 99%
Synonym(s):
(+)-2-Hydroxyisopinocampheol, (1S,2S,3R,5S)-2,3-Pinanediol, (1S,2S,3R,5S)-2,6,6-Trimethylbicyclo[3.1.1]heptane-2,3-diol, [1S-(1α,2α,3α,5α)]-2,6,6-Trimethylbicyclo[3.1.1]heptane-2,3-diol
Empirical Formula (Hill Notation):
C10H18O2
CAS Number:
Molecular Weight:
170.25

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing