Skip to Content
MilliporeSigma
(1R)-(−)-Camphorquinone 99%

(1R)-(−)-Camphorquinone

(1R)-(−)-Camphorquinone 99%
Synonym(s):
(1R)-(−)-2,3-Bornanedione, 2,3-Bornanedione
Empirical Formula (Hill Notation):
C10H14O2
CAS Number:
Molecular Weight:
166.22

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing