Skip to Content
MilliporeSigma
(1R,2R)-(−)-1,2-Diaminocyclohexane 98%

(1R,2R)-(−)-1,2-Diaminocyclohexane

(1R,2R)-(−)-1,2-Diaminocyclohexane 98%
Synonym(s):
(1R)-trans-1,2-Cyclohexanediamine
Linear Formula:
C6H10(NH2)2
CAS Number:
Molecular Weight:
114.19

Products

Product Category
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing