Skip to Content
MilliporeSigma
(1R)-(−)-10-Camphorsulfonic acid ammonium salt 98%

(1R)-(−)-10-Camphorsulfonic acid ammonium salt

(1R)-(−)-10-Camphorsulfonic acid ammonium salt 98%
Empirical Formula (Hill Notation):
C10H19NO4S
CAS Number:
Molecular Weight:
249.33
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing