Skip to Content
MilliporeSigma
(1R)-(−)-10-Camphorsulfonic acid 98%

(1R)-(−)-10-Camphorsulfonic acid

(1R)-(−)-10-Camphorsulfonic acid 98%
Synonym(s):
(−)-Camphor-10-sulfonic acid, (1R)-(−)-Camphor-10-sulfonic acid, (1R)-Camphor-10-sulfonic acid
Empirical Formula (Hill Notation):
C10H16O4S
CAS Number:
Molecular Weight:
232.30
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing