Skip to Content
MilliporeSigma
1-Phenyl-2-trimethylsilylacetylene 99%

1-Phenyl-2-trimethylsilylacetylene

1-Phenyl-2-trimethylsilylacetylene 99%
Synonym(s):
1-(Trimethylsilyl)-2-phenylacetylene, Phenylethynyl-trimethylsilane, Trimethyl(phenylethynyl)silane
Linear Formula:
C6H5C≡CSi(CH3)3
CAS Number:
Molecular Weight:
174.31

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing