Skip to Content
MilliporeSigma
1-Octanethiol ≥98.5%

1-Octanethiol

1-Octanethiol ≥98.5%
Synonym(s):
n-Octyl mercaptan, Mercaptan C8, Octyl mercaptan, 1-Octanethiol
Linear Formula:
CH3(CH2)7SH
CAS Number:
Molecular Weight:
146.29

Products

Brand
Boiling Point (°C)
Formula Weight
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing