Skip to Content
MilliporeSigma
1-Methyl-L-tryptophan 95%

1-Methyl-L-tryptophan

1-Methyl-L-tryptophan 95%
Empirical Formula (Hill Notation):
C12H14N2O2
CAS Number:
Molecular Weight:
218.25

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing