Skip to Content
MilliporeSigma
1-Hydroxypyridine-2-thione zinc salt ≥95%

1-Hydroxypyridine-2-thione zinc salt

1-Hydroxypyridine-2-thione zinc salt ≥95%
Synonym(s):
Mercaptopyridine N-oxide zinc salt, Pyrithione
Empirical Formula (Hill Notation):
C10H8N2O2S2Zn
CAS Number:
Molecular Weight:
317.70
EC Number:

Products

Product Category
Brand
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing