Skip to Content
MilliporeSigma
1-Hexyl-3-methylimidazolium chloride ≥97.0% (HPLC)

1-Hexyl-3-methylimidazolium chloride

1-Hexyl-3-methylimidazolium chloride ≥97.0% (HPLC)
Synonym(s):
HMIMCl
Empirical Formula (Hill Notation):
C10H19ClN2
CAS Number:
Molecular Weight:
202.72

Products

Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing