Skip to Content
MilliporeSigma
1-Hexyl-3-methylimidazolium bis(trifluormethylsulfonyl)imide 98%

1-Hexyl-3-methylimidazolium bis(trifluormethylsulfonyl)imide

1-Hexyl-3-methylimidazolium bis(trifluormethylsulfonyl)imide 98%
Synonym(s):
HMIM BTI, HMIM TFS
Empirical Formula (Hill Notation):
C12H19F6N3O4S2
CAS Number:
Molecular Weight:
447.42

Products

Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing