Skip to Content
MilliporeSigma
1H,1H,2H-Perfluoro-1-decene 99%

1H,1H,2H-Perfluoro-1-decene

1H,1H,2H-Perfluoro-1-decene 99%
Synonym(s):
3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluoro-1-decene
Linear Formula:
CF3(CF2)7CH=CH2
CAS Number:
Molecular Weight:
446.10
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing