Skip to Content
MilliporeSigma
1H,1H,2H,2H-Perfluorodecanethiol 97%

1H,1H,2H,2H-Perfluorodecanethiol

1H,1H,2H,2H-Perfluorodecanethiol 97%
Synonym(s):
3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluoro-1-decanethiol, 1H,1H,2H,2H-Perfluoro-1-decanethiol
Linear Formula:
CF3(CF2)7CH2CH2SH
CAS Number:
Molecular Weight:
480.18

Products

Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing