Skip to Content
MilliporeSigma
1-Butyl-3-methylimidazolium tetrafluoroborate for catalysis, ≥98.5% (HPLC)

1-Butyl-3-methylimidazolium tetrafluoroborate

1-Butyl-3-methylimidazolium tetrafluoroborate for catalysis, ≥98.5% (HPLC)
Synonym(s):
BMIMBF4
Empirical Formula (Hill Notation):
C8H15BF4N2
CAS Number:
Molecular Weight:
226.02

Products

Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group