Skip to Content
MilliporeSigma
1-Butyl-3-methylimidazolium octyl sulfate ≥95.0% (HPLC)

1-Butyl-3-methylimidazolium octyl sulfate

1-Butyl-3-methylimidazolium octyl sulfate ≥95.0% (HPLC)
Synonym(s):
BMIM OSU
Empirical Formula (Hill Notation):
C16H32N2O4S
CAS Number:
Molecular Weight:
348.50

Products

Formula Weight
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing