Skip to Content
MilliporeSigma
1-Butyl-3-methylimidazolium chloride ≥99.0% (HPLC)

1-Butyl-3-methylimidazolium chloride

1-Butyl-3-methylimidazolium chloride ≥99.0% (HPLC)
Synonym(s):
BMIMCl
Empirical Formula (Hill Notation):
C8H15ClN2
CAS Number:
Molecular Weight:
174.67

Products

Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Special Grade