Skip to Content
MilliporeSigma
1-Butyl-3-methylimidazolium bis(trifluoromethylsulfonyl)imide ≥99%, H2O <500 ppm

1-Butyl-3-methylimidazolium bis(trifluoromethylsulfonyl)imide

1-Butyl-3-methylimidazolium bis(trifluoromethylsulfonyl)imide ≥99%, H2O <500 ppm
Synonym(s):
BMIIm, BMIM TFSI
Empirical Formula (Hill Notation):
C10H15F6N3O4S2
CAS Number:
Molecular Weight:
419.36

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Chemical Composition
Quality Segment
Application
Markush Class
Markush Group
Special Grade