Skip to Content
MilliporeSigma
1-Butyl-3-methylimidazolium acetate ≥96.0% (HPLC)

1-Butyl-3-methylimidazolium acetate

1-Butyl-3-methylimidazolium acetate ≥96.0% (HPLC)
Empirical Formula (Hill Notation):
C10H18N2O2
CAS Number:
Molecular Weight:
198.26

Products

Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing