Skip to Content
MilliporeSigma
1-Butyl-2,3-dimethylimidazolium chloride ≥97.0% (HPLC/AT)

1-Butyl-2,3-dimethylimidazolium chloride

1-Butyl-2,3-dimethylimidazolium chloride ≥97.0% (HPLC/AT)
Empirical Formula (Hill Notation):
C9H17ClN2
CAS Number:
Molecular Weight:
188.70

Products

Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing