Skip to Content
MilliporeSigma
1-Butyl-1-methylpiperidinium bis(trifluoromethylsulfonyl)imide 97%

1-Butyl-1-methylpiperidinium bis(trifluoromethylsulfonyl)imide

1-Butyl-1-methylpiperidinium bis(trifluoromethylsulfonyl)imide 97%
Empirical Formula (Hill Notation):
C12H22F6N2O4S2
CAS Number:
Molecular Weight:
436.43

Products

Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing