Skip to Content
MilliporeSigma
1-Bromobutane analytical standard

1-Bromobutane

1-Bromobutane analytical standard
Synonym(s):
1-Bromobutane, Butyl bromide
Linear Formula:
CH3(CH2)3Br
CAS Number:
Molecular Weight:
137.02
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing