Skip to Content
MilliporeSigma
1-Bromo-2,4,6-tri-tert-butylbenzene 97%

1-Bromo-2,4,6-tri-tert-butylbenzene

1-Bromo-2,4,6-tri-tert-butylbenzene 97%
Linear Formula:
[(CH3)3C]3C6H2Br
CAS Number:
Molecular Weight:
325.33

Products

Product Category
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing