Skip to Content
MilliporeSigma
1-Boc-4-piperidone 98%

1-Boc-4-piperidone

1-Boc-4-piperidone 98%
Synonym(s):
N-Boc-4-piperidone, tert-Butyl 4-oxo-1-piperidinecarboxylate
Empirical Formula (Hill Notation):
C10H17NO3
CAS Number:
Molecular Weight:
199.25

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing