Skip to Content
MilliporeSigma
1-Boc-4-hydroxypiperidine 97%

1-Boc-4-hydroxypiperidine

1-Boc-4-hydroxypiperidine 97%
Synonym(s):
1-Boc-4-piperidinol, tert-Butyl 4-hydroxy-1-piperidinecarboxylate
Empirical Formula (Hill Notation):
C10H19NO3
CAS Number:
Molecular Weight:
201.26

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing