Skip to Content
MilliporeSigma
1-Boc-3-azetidinone 97%

1-Boc-3-azetidinone

1-Boc-3-azetidinone 97%
Synonym(s):
tert-Butyl-3-oxoazetidine-1-carboxylate
Empirical Formula (Hill Notation):
C8H13NO3
CAS Number:
Molecular Weight:
171.19

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing