Skip to Content
MilliporeSigma
1-Azido-1-deoxy-β-D-glucopyranoside

1-Azido-1-deoxy-β-D-glucopyranoside

1-Azido-1-deoxy-β-D-glucopyranoside
Empirical Formula (Hill Notation):
C6H11N3O5
CAS Number:
Molecular Weight:
205.17

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Markush Class
Markush Group
Optical Isomer
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing