Skip to Content
MilliporeSigma
1α-Hydroxyvitamin D3 ≥97.0% (HPLC)

1α-Hydroxyvitamin D3

1α-Hydroxyvitamin D3 ≥97.0% (HPLC)
Synonym(s):
1α-Hydroxycholecalciferol, Alfacalcidol
Empirical Formula (Hill Notation):
C27H44O2
CAS Number:
Molecular Weight:
400.64
EC Number:

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing