![1α,25-Dihydroxyvitamin D2 solution 100 μg/mL in ethanol, 95% (CP)](/deepweb/assets/sigmaaldrich/product/structures/424/837/797bfa7d-8cb2-492e-84d0-11cdf290b1b9/640/797bfa7d-8cb2-492e-84d0-11cdf290b1b9.png)
1α,25-Dihydroxyvitamin D2 solution
![1α,25-Dihydroxyvitamin D2 solution 100 μg/mL in ethanol, 95% (CP)](/deepweb/assets/sigmaaldrich/product/structures/424/837/797bfa7d-8cb2-492e-84d0-11cdf290b1b9/640/797bfa7d-8cb2-492e-84d0-11cdf290b1b9.png)
Synonym(s):
1α,25-Dihydroxycalciferol solution
Empirical Formula (Hill Notation):
C28H44O3
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing