Skip to Content
MilliporeSigma
1,8-Diiodooctane 98%, contains copper as stabilizer

1,8-Diiodooctane

1,8-Diiodooctane 98%, contains copper as stabilizer
Synonym(s):
Octamethylene diiodide
Linear Formula:
I(CH2)8I
CAS Number:
Molecular Weight:
366.02
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing