Skip to Content
MilliporeSigma
1,8-Dihydroxynaphthalene 95%

1,8-Dihydroxynaphthalene

1,8-Dihydroxynaphthalene 95%
Synonym(s):
1,8-Naphthalenediol
Empirical Formula (Hill Notation):
C10H8O2
CAS Number:
Molecular Weight:
160.17

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing