Skip to Content
MilliporeSigma
1,6-Dibromohexane 96%

1,6-Dibromohexane

1,6-Dibromohexane 96%
Synonym(s):
Hexamethylene dibromide, 1,6-Dibromohexane
Linear Formula:
Br(CH2)6Br
CAS Number:
Molecular Weight:
243.97

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing