Skip to Content
MilliporeSigma
16-Bromohexadecanoic acid ≥99%

16-Bromohexadecanoic acid

16-Bromohexadecanoic acid ≥99%
Linear Formula:
BrCH2(CH2)14CO2H
CAS Number:
Molecular Weight:
335.32

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing