Skip to Content
MilliporeSigma
(16a)-16,17,21-Trihydroxypregna-1,4,9(11)-triene-3,20-dione Pharmaceutical Analytical Impurity (PAI)

(16a)-16,17,21-Trihydroxypregna-1,4,9(11)-triene-3,20-dione

(16a)-16,17,21-Trihydroxypregna-1,4,9(11)-triene-3,20-dione Pharmaceutical Analytical Impurity (PAI)
Synonym(s):
(16α)-16,17,21-Trihydroxypregna-1,4,9(11)-triene-3,20-dione, 16α,17,21-Trihydroxy-pregna-1,4,9(11)-triene-3,20-dione
Empirical Formula (Hill Notation):
C21H26O5
CAS Number:
Molecular Weight:
358.43

Products

Formula Weight
Manufacturer
Agency
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1