Skip to Content
MilliporeSigma
1,4-Pentadien-3-ol contains 0.4% hydroquinone as stabilizer, ≥96%

1,4-Pentadien-3-ol

1,4-Pentadien-3-ol contains 0.4% hydroquinone as stabilizer, ≥96%
Synonym(s):
Divinyl carbinol
Linear Formula:
CH2=CHCH(OH)CH=CH2
CAS Number:
Molecular Weight:
84.12

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing