![14-Deoxy-11,12-didehydroandrographolide analytical standard](/deepweb/assets/sigmaaldrich/product/structures/699/080/8ad8fe4a-73ed-467e-bbdc-a4a08225cc51/640/8ad8fe4a-73ed-467e-bbdc-a4a08225cc51.png)
14-Deoxy-11,12-didehydroandrographolide
![14-Deoxy-11,12-didehydroandrographolide analytical standard](/deepweb/assets/sigmaaldrich/product/structures/699/080/8ad8fe4a-73ed-467e-bbdc-a4a08225cc51/640/8ad8fe4a-73ed-467e-bbdc-a4a08225cc51.png)
Synonym(s):
3-[(1E)-2-[(1R,4aS,5R,6R,8aR)-Decahydro-6-hydroxy-5-(hydroxymethyl)-5,8a-dimethyl-2-methylene-1-naphthalenyl]ethenyl]- 2(5H)-furanone, ent-(3β,11E)-3,19-Dihydroxy-8(17),11,13-labdatrien-16,15-olide
Empirical Formula (Hill Notation):
C20H28O4
CAS Number:
Molecular Weight:
332.43
Products
Brand
Formula Weight
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing