Skip to Content
MilliporeSigma
1,4-Bis(trimethylsilyl)benzene 96%

1,4-Bis(trimethylsilyl)benzene

1,4-Bis(trimethylsilyl)benzene 96%
Synonym(s):
p-Phenylenebis(trimethylsilane)
Linear Formula:
C6H4[Si(CH3)3]2
CAS Number:
Molecular Weight:
222.47

Products

Product Category
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing