Skip to Content
MilliporeSigma
1,4-Benzodioxane-6-acetic acid 97%

1,4-Benzodioxane-6-acetic acid

1,4-Benzodioxane-6-acetic acid 97%
Empirical Formula (Hill Notation):
C10H10O4
CAS Number:
Molecular Weight:
194.18
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing