Skip to Content
MilliporeSigma
1,4,8,11-Tetramethyl-1,4,8,11-tetraazacyclotetradecane 98%

1,4,8,11-Tetramethyl-1,4,8,11-tetraazacyclotetradecane

1,4,8,11-Tetramethyl-1,4,8,11-tetraazacyclotetradecane 98%
Empirical Formula (Hill Notation):
C14H32N4
CAS Number:
Molecular Weight:
256.43

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing