Skip to Content
MilliporeSigma
1,3-Butadiene diepoxide 97%

1,3-Butadiene diepoxide

1,3-Butadiene diepoxide 97%
Synonym(s):
1,2,3,4-Diepoxybutane
Empirical Formula (Hill Notation):
C4H6O2
CAS Number:
Molecular Weight:
86.09

Products

Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing